Product Details

SNP ID
rs1697946
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.5:619538 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCGATGCCTGAGCATTATGGAGGC[C/T]GCGTCTGCATGGCCTGATTGGCCCG
Phenotype
MIM: 616475
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
CEP72 PubMed Links

Gene Details

Gene
CEP72
Gene Name
centrosomal protein 72
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018140.3 Intron NP_060610.2
XM_005248322.3 Intron XP_005248379.1
XM_011514063.1 Intron XP_011512365.1
XM_011514064.2 Intron XP_011512366.1
XM_017009626.1 Intron XP_016865115.1
XM_017009627.1 Intron XP_016865116.1
Gene
LOC100996325
Gene Name
uncharacterized LOC100996325
There are no transcripts associated with this gene.

View Full Product Details