Product Details

SNP ID
rs1949972
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:80745122 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CCCTCACAGAGCTGGCCCAGGTCCA[C/T]GGGGACCATTTGTCCGTCACAACAG
Phenotype
MIM: 602645
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
SEMA3C PubMed Links

Gene Details

Gene
SEMA3C
Gene Name
semaphorin 3C
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006379.3 2590 Silent Mutation CCA,CCG P676P NP_006370.1
XM_005250113.2 2590 Silent Mutation CCA,CCG P618P XP_005250170.1

View Full Product Details