Product Details

SNP ID
rs1133950
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:116771675 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AAACACTGAACAGAGTAGAAGAAAA[A/C]AGCGCAAGAAAATAAGTGGTCCCAA
Phenotype
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
UTP23 PubMed Links

Gene Details

Gene
UTP23
Gene Name
UTP23, small subunit processome component
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_032334.2 684 Missense Mutation AAG,CAG K195Q NP_115710.2

View Full Product Details