Product Details

SNP ID
rs7224977
Assay Type
Functionally Tested
NCBI dbSNP Submissions
NA
Location
Chr.17:1781478 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGTAAGAGGAGTTAGTTCACTGATT[C/T]GTGAGGCAAATGTTAATTGAGGGCC
Phenotype
MIM: 172860
Polymorphism
C/T, Transition Substitution
Allele Nomenclature
Literature Links
SERPINF1 PubMed Links

Gene Details

Gene
SERPINF1
Gene Name
serpin family F member 1
There are no transcripts associated with this gene.

Gene
SMYD4
Gene Name
SET and MYND domain containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_052928.2 Intron NP_443160.2
XM_011523632.2 Intron XP_011521934.1
XM_011523633.2 Intron XP_011521935.1
XM_017024121.1 Intron XP_016879610.1

View Full Product Details