Product Details

SNP ID
rs15943
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.X:19361522 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TCAATTGTCTTTGCATCAGCTCCTT[C/G]CAGCCGCAACCAGTCTATAAGCTCT
Phenotype
MIM: 300820 MIM: 300502
Polymorphism
C/G, Transversion substitution
Allele Nomenclature
Literature Links
MAP3K15 PubMed Links

Gene Details

Gene
MAP3K15
Gene Name
mitogen-activated protein kinase kinase kinase 15
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001001671.3 3301 Missense Mutation CAA,GAA Q1251E NP_001001671.3
XM_011545507.2 3301 Missense Mutation CAA,GAA Q1262E XP_011543809.2
XM_011545508.2 3301 Missense Mutation CAA,GAA Q1233E XP_011543810.2
XM_011545510.2 3301 Missense Mutation CAA,GAA Q809E XP_011543812.1
XM_011545511.2 3301 Missense Mutation CAA,GAA Q686E XP_011543813.1
XM_017029511.1 3301 Intron XP_016885000.1
Gene
PDHA1
Gene Name
pyruvate dehydrogenase (lipoamide) alpha 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000284.3 3301 UTR 3 NP_000275.1
NM_001173454.1 3301 UTR 3 NP_001166925.1
NM_001173455.1 3301 UTR 3 NP_001166926.1
NM_001173456.1 3301 UTR 3 NP_001166927.1
XM_017029574.1 3301 Intron XP_016885063.1

View Full Product Details