Product Details

SNP ID
rs2231312
Assay Type
Functionally tested
NCBI dbSNP Submissions
43
Location
Chr.1:36139877 on Build GRCh38
Set Membership
HapMap JSNP
Context Sequence [VIC/FAM]
GAGTCTAAATGTCTTATGTTTGAAC[A/G]TAAGGTTGTTGGTGGTAAAGGGAAA
Phenotype
MIM: 610955
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
TRAPPC3 PubMed Links

Gene Details

Gene
TRAPPC3
Gene Name
trafficking protein particle complex 3
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270894.1 Intron NP_001257823.1
NM_001270895.1 Intron NP_001257824.1
NM_001270896.1 Intron NP_001257825.1
NM_001270897.1 Intron NP_001257826.1
NM_014408.4 Intron NP_055223.1

View Full Product Details