Product Details
- SNP ID
-
rs2299742
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.21:39868098 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- GCGGCCCCGGCAGAAGAAGAGCTAC[A/G]ATTTTGCAAATCCAAAGTCACTTCT
- Phenotype
-
MIM: 601629
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PCP4
PubMed Links
Gene Details
- Gene
- PCP4
- Gene Name
- Purkinje cell protein 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_006198.2 |
|
Intron |
|
|
NP_006189.2 |
View Full Product Details