Product Details

SNP ID
rs2300596
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.14:74887152 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AATATAAATATTATCAGATTTATCT[C/T]AAAATGGCAAAGAGCAAGGGATTGG
Phenotype
MIM: 126063
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DLST PubMed Links

Gene Details

Gene
DLST
Gene Name
dihydrolipoamide S-succinyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001244883.1 Intron NP_001231812.1
NM_001933.4 Intron NP_001924.2

View Full Product Details