Product Details

SNP ID
rs2301614
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.9:133460038 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GGCCGGCTACCGAGCCCTTTGTGAG[A/G]GCTGTGAGCTGCGCCTGACGGTGGC
Phenotype
MIM: 604134 MIM: 613104
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
ADAMTS13 PubMed Links

Gene Details

Gene
ADAMTS13
Gene Name
ADAM metallopeptidase with thrombospondin type 1 motif 13
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_139025.4 74 Intron NP_620594.1
NM_139026.4 74 Intron NP_620595.1
NM_139027.4 74 Intron NP_620596.2
XM_011518176.2 74 Intron XP_011516478.1
XM_011518178.1 74 Intron XP_011516480.1
XM_011518179.1 74 Intron XP_011516481.1
XM_017014232.1 74 Intron XP_016869721.1
XM_017014233.1 74 Intron XP_016869722.1
XM_017014234.1 74 Intron XP_016869723.1
XM_017014235.1 74 Intron XP_016869724.1
Gene
CACFD1
Gene Name
calcium channel flower domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001135775.2 74 UTR 5 NP_001129247.1
NM_001242369.1 74 UTR 5 NP_001229298.1
NM_001242370.1 74 UTR 5 NP_001229299.1
NM_017586.3 74 UTR 5 NP_060056.1
XM_011518181.2 74 Intron XP_011516483.1
XM_011518182.1 74 UTR 5 XP_011516484.1
XM_011518184.1 74 Intron XP_011516486.1
XM_011518185.2 74 Intron XP_011516487.1
XM_011518186.1 74 UTR 5 XP_011516488.1

View Full Product Details