Product Details

SNP ID
rs2233474
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:50351176 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTGTAGTCTTACATGGCTTTGCGC[A/C]TCCTACCTGGAAGGCGGGCCAGCGA
Phenotype
MIM: 607068 MIM: 607072 MIM: 607069 MIM: 607070
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
CYB561D2 PubMed Links

Gene Details

Gene
CYB561D2
Gene Name
cytochrome b561 family member D2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291284.1 Intron NP_001278213.1
NM_007022.4 Intron NP_008953.1
Gene
NPRL2
Gene Name
NPR2-like, GATOR1 complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006545.4 Intron NP_006536.3
XM_005264808.4 Intron XP_005264865.1
XM_011533288.2 Intron XP_011531590.1
XM_017005555.1 Intron XP_016861044.1
XM_017005556.1 Intron XP_016861045.1
Gene
TMEM115
Gene Name
transmembrane protein 115
There are no transcripts associated with this gene.

Gene
ZMYND10
Gene Name
zinc finger MYND-type containing 10
There are no transcripts associated with this gene.

View Full Product Details