Product Details

SNP ID
rs3009184
Assay Type
Validated
NCBI dbSNP Submissions
46
Location
Chr.1:119837385 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
CTGTAAACAAATTCAGAAGAGCAGG[A/T]CATATTATGCAAATCACGCTTCACA
Phenotype
MIM: 613997
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
NBPF7 PubMed Links

Gene Details

Gene
NBPF7
Gene Name
neuroblastoma breakpoint family member 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001047980.2 Intron NP_001041445.1
XM_017001186.1 Intron XP_016856675.1

View Full Product Details