Product Details
- SNP ID
-
rs2302982
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.5:98771628 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GGCTTATAAATCTCGAGCTTGTCAA[A/C]TTTTACAACAGAGGAGCAATAAAGC
- Phenotype
-
MIM: 612687
- Polymorphism
- A/C, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
RGMB
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs79139926] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- RGMB
- Gene Name
- repulsive guidance molecule family member b
- Gene
- RGMB-AS1
- Gene Name
- RGMB antisense RNA 1
There are no transcripts associated with this gene.
View Full Product Details