Product Details
- SNP ID
-
rs199742451
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.4:4188849 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- AAAGAGGGAGGCAGCTGCGTGCATT[C/G]GATAGAAAATAGAAAAAGGCATGGC
- Phenotype
-
MIM: 607806
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
OTOP1
PubMed Links
Gene Details
- Gene
- OTOP1
- Gene Name
- otopetrin 1
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_177998.1 |
1823 |
Missense Mutation |
|
|
NP_819056.1 |
View Full Product Details