Product Details

SNP ID
rs2344745
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:170296477 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGGCGGCGCCAGGGCCCTCCCGGTG[A/G]ACCCCAGTCGCCGTCGTGACGGCCC
Phenotype
MIM: 606582 MIM: 612266
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
DLL1 PubMed Links

Gene Details

Gene
DLL1
Gene Name
delta like canonical Notch ligand 1
There are no transcripts associated with this gene.

Gene
FAM120B
Gene Name
family with sequence similarity 120B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001286379.1 Intron NP_001273308.1
NM_001286380.1 Intron NP_001273309.1
NM_001286381.1 Intron NP_001273310.1
NM_032448.2 Intron NP_115824.1
XM_011536185.2 Intron XP_011534487.1
XM_017011357.1 Intron XP_016866846.1
XM_017011358.1 Intron XP_016866847.1
XM_017011359.1 Intron XP_016866848.1
XM_017011360.1 Intron XP_016866849.1

View Full Product Details