Product Details

SNP ID
rs2069772
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:122451978 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTTATTTTCAAGCTTACCAAACATA[C/T]TAATTATTCACATTTTCTTGAAATG
Phenotype
MIM: 147680
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL2 PubMed Links

Gene Details

Gene
IL2
Gene Name
interleukin 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000586.3 Intron NP_000577.2
XM_017008177.1 Intron XP_016863666.1

View Full Product Details