Product Details

SNP ID
rs3087570
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.6:33250768 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAAAACTGGAAGGGGTACCCCAGG[G/T]GAAGAAGGGTATGGAATGGGGTGCA
Phenotype
MIM: 603443
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
HCG25 PubMed Links

Gene Details

Gene
HCG25
Gene Name
HLA complex group 25 (non-protein coding)
There are no transcripts associated with this gene.

Gene
VPS52
Gene Name
VPS52, GARP complex subunit
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001289174.1 2414 UTR 3 NP_001276103.1
NM_001289175.1 2414 UTR 3 NP_001276104.1
NM_001289176.1 2414 UTR 3 NP_001276105.1
NM_022553.5 2414 UTR 3 NP_072047.4
XM_011514797.1 2414 UTR 3 XP_011513099.1
XM_011514798.2 2414 UTR 3 XP_011513100.1
XM_011514799.1 2414 UTR 3 XP_011513101.1
XM_017011177.1 2414 Intron XP_016866666.1
XM_017011178.1 2414 UTR 3 XP_016866667.1
XM_017011179.1 2414 Intron XP_016866668.1

View Full Product Details