Product Details

SNP ID
hCV15802611
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.16:15070589 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCAAACAGGACACGTTCACCAACAC[G/C]TCTCAAGAGAAAGACAGTCTGGTGG
Phenotype
MIM: 614244 MIM: 605121
Polymorphism
G/C, Transversion substitution
Allele Nomenclature
Literature Links
PDXDC1 PubMed Links

Gene Details

Gene
PDXDC1
Gene Name
pyridoxal dependent decarboxylase domain containing 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285444.1 Intron NP_001272373.1
NM_001285445.1 Intron NP_001272374.1
NM_001285447.1 Intron NP_001272376.1
NM_001285448.1 Intron NP_001272377.1
NM_001285449.1 Intron NP_001272378.1
NM_001285450.1 Intron NP_001272379.1
NM_001324019.1 Intron NP_001310948.1
NM_001324020.1 Intron NP_001310949.1
NM_001324021.1 Intron NP_001310950.1
NM_015027.3 Intron NP_055842.2
Gene
RRN3
Gene Name
RRN3 homolog, RNA polymerase I transcription factor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001301064.1 Intron NP_001287993.1
NM_018427.4 Intron NP_060897.3

View Full Product Details