Product Details
- SNP ID
-
rs2842946
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.6:18131602 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TCTCAAAAATAAACAAATAAAAAAA[A/T]ATATACCATCTCTCTAGAAAAGTTC
- Phenotype
-
MIM: 608072
MIM: 187680
- Polymorphism
- A/T, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
NHLRC1
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114611199] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- NHLRC1
- Gene Name
- NHL repeat containing E3 ubiquitin protein ligase 1
There are no transcripts associated with this gene.
- Gene
- TPMT
- Gene Name
- thiopurine S-methyltransferase
View Full Product Details