Product Details
- SNP ID
-
rs2140423
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.3:187724380 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCTGCCGAGACCCCGCCCTAGAACA[C/T]TCCAATCCCAGTGAACCTATCCACA
- Phenotype
-
MIM: 109565
MIM: 609138
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
BCL6
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs114571972] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- BCL6
- Gene Name
- B-cell CLL/lymphoma 6
- Gene
- LOC100131635
- Gene Name
- hCG1645011-like
There are no transcripts associated with this gene.
- Gene
- RTP2
- Gene Name
- receptor transporter protein 2
There are no transcripts associated with this gene.
View Full Product Details