Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001017969.2 | Intron | NP_001017969.2 | |||
XM_005251385.2 | Intron | XP_005251442.1 | |||
XM_006716730.3 | Intron | XP_006716793.1 | |||
XM_011517759.2 | Intron | XP_011516061.1 | |||
XM_011517760.2 | Intron | XP_011516062.1 | |||
XM_011517761.2 | Intron | XP_011516063.1 | |||
XM_017014331.1 | Intron | XP_016869820.1 | |||
XM_017014332.1 | Intron | XP_016869821.1 | |||
XM_017014333.1 | Intron | XP_016869822.1 | |||
XM_017014334.1 | Intron | XP_016869823.1 | |||
XM_017014335.1 | Intron | XP_016869824.1 | |||
XM_017014336.1 | Intron | XP_016869825.1 |
Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_005511.1 | Intron | NP_005502.1 |