Product Details

SNP ID
rs2862139
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.15:99261445 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGTTGGGGAAGTACAAGATCAGGGT[A/G]CTAGCAGATTCGGTGTCTGGTGAGG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
HSP90B2P PubMed Links

Gene Details

Gene
HSP90B2P
Gene Name
heat shock protein 90 beta family member 2, pseudogene
There are no transcripts associated with this gene.

Gene
LRRC28
Gene Name
leucine rich repeat containing 28
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001284400.2 Intron NP_001271329.1
NM_001321675.1 Intron NP_001308604.1
NM_001321676.1 Intron NP_001308605.1
NM_001321677.1 Intron NP_001308606.1
NM_001321678.1 Intron NP_001308607.1
NM_001321679.1 Intron NP_001308608.1
NM_001321680.1 Intron NP_001308609.1
NM_144598.4 Intron NP_653199.2
XM_006720389.3 Intron XP_006720452.1
XM_011521218.2 Intron XP_011519520.1
XM_011521220.1 Intron XP_011519522.1
XM_011521221.2 Intron XP_011519523.1
XM_017021914.1 Intron XP_016877403.1

View Full Product Details