Product Details

SNP ID
rs2862852
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.2:113059019 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GCCTCAAGGAAGTTCTTCAATATTT[C/T]TGGAAGGAAACTGTCTCCAGAAGCT
Phenotype
MIM: 615296 MIM: 605508 MIM: 605507
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
IL1F10 PubMed Links

Gene Details

Gene
IL1F10
Gene Name
interleukin 1 family member 10 (theta)
There are no transcripts associated with this gene.

Gene
IL36B
Gene Name
interleukin 36, beta
There are no transcripts associated with this gene.

Gene
IL36RN
Gene Name
interleukin 36 receptor antagonist
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_012275.2 Intron NP_036407.1
NM_173170.1 Intron NP_775262.1

View Full Product Details