Product Details

SNP ID
rs2179208
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.X:46578023 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
GGCCAGGATTTGGCAGTGAGGGAGT[C/T]AGTGAAGTTTTTTCTGGTCTGGGTA
Phenotype
MIM: 300375
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
CHST7 PubMed Links

Gene Details

Gene
CHST7
Gene Name
carbohydrate sulfotransferase 7
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_019886.3 Intron NP_063939.2

View Full Product Details