Product Details

SNP ID
rs2518799
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:18984912 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AGGAGGAATTCTGATATAACAGGGA[C/T]GTCTCCACATGTCTTCCCTGCCTTA
Phenotype
Polymorphism
C/T, Transition substitution
Allele Nomenclature
Literature Links
DGCR5 PubMed Links

Gene Details

Gene
DGCR5
Gene Name
DiGeorge syndrome critical region gene 5 (non-protein coding)
There are no transcripts associated with this gene.

View Full Product Details