Product Details

SNP ID
rs2886630
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.17:70075786 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTATGAACATATAATTGCAAATATT[A/G]AAAAATATGTATTTTATACCCCCCT
Phenotype
MIM: 605722
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
KCNJ16 PubMed Links

Gene Details

Gene
KCNJ16
Gene Name
potassium voltage-gated channel subfamily J member 16
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001270422.1 Intron NP_001257351.1
NM_001291622.1 Intron NP_001278551.1
NM_001291623.1 Intron NP_001278552.1
NM_001291624.1 Intron NP_001278553.1
NM_001291625.1 Intron NP_001278554.1
NM_018658.2 Intron NP_061128.2
NM_170741.2 Intron NP_733937.2
NM_170742.2 Intron NP_733938.2
XM_005257337.4 Intron XP_005257394.1
XM_006721885.3 Intron XP_006721948.1
XM_006721886.3 Intron XP_006721949.1
XM_006721887.3 Intron XP_006721950.1
XM_011524781.2 Intron XP_011523083.1
XM_017024609.1 Intron XP_016880098.1
XM_017024610.1 Intron XP_016880099.1
XM_017024611.1 Intron XP_016880100.1
XM_017024612.1 Intron XP_016880101.1
XM_017024613.1 Intron XP_016880102.1
Gene
LINC01028
Gene Name
long intergenic non-protein coding RNA 1028
There are no transcripts associated with this gene.

View Full Product Details