Product Details

SNP ID
rs2180403
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.22:49864018 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TTCTGTGCTCATCTTGTATTTTTCC[G/T]GCTCCAGACCTGAAATAAGCCATTT
Phenotype
MIM: 607144 MIM: 612552
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
ALG12 PubMed Links

Gene Details

Gene
ALG12
Gene Name
ALG12, alpha-1,6-mannosyltransferase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_024105.3 Intron NP_077010.1
XM_011530371.2 Intron XP_011528673.1
XM_017028936.1 Intron XP_016884425.1
XM_017028937.1 Intron XP_016884426.1
Gene
ZBED4
Gene Name
zinc finger BED-type containing 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_014838.2 Intron NP_055653.2

View Full Product Details