Product Details

SNP ID
rs2187566
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68156171 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATGGGCTACTTTCCTATAGACACTA[A/T]ACAAATCCCCTGAATTGATTTTTAC
Phenotype
MIM: 610881
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
KMT5B PubMed Links

Gene Details

Gene
KMT5B
Gene Name
lysine methyltransferase 5B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001300907.1 4527 UTR 3 NP_001287836.1
NM_001300908.1 4527 UTR 3 NP_001287837.1
NM_001300909.1 4527 Intron NP_001287838.1
NM_016028.4 4527 Intron NP_057112.3
NM_017635.4 4527 UTR 3 NP_060105.3

View Full Product Details