Product Details
- SNP ID
-
rs2923131
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.16:57695664 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- GGGGTCCTGAATGGTGTTGGGCTAC[C/T]TAGGCCTTGGGTGGCTTCCAGCCAG
- Phenotype
-
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ADGRG3
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs77257519] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ADGRG3
- Gene Name
- adhesion G protein-coupled receptor G3
There are no transcripts associated with this gene.
- Gene
- DRC7
- Gene Name
- dynein regulatory complex subunit 7
View Full Product Details