Product Details

SNP ID
rs2232017
Assay Type
Functionally tested
NCBI dbSNP Submissions
22
Location
Chr.1:107057657 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TGCTGTCCACCTCGCCTTTTCACCC[A/G]GCCACTCACTGGAAACAGGCGCTCC
Phenotype
MIM: 608274
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
PRMT6 PubMed Links

Gene Details

Gene
PRMT6
Gene Name
protein arginine methyltransferase 6
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_018137.2 1013 Silent Mutation CCA,CCG P314P NP_060607.2

View Full Product Details