Product Details

SNP ID
rs2924701
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:68904620 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
ATTGGAGTTAAGCTGGTGGGTGCGG[A/G]GTAGGGTGGGGAGCCGTCCTGAGAG
Phenotype
MIM: 600502 MIM: 611834
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IGHMBP2 PubMed Links

Gene Details

Gene
IGHMBP2
Gene Name
immunoglobulin mu binding protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_002180.2 Intron NP_002171.2
XM_005273974.2 Intron XP_005274031.1
XM_005273975.3 Intron XP_005274032.1
XM_005273976.1 Intron XP_005274033.1
XM_011544994.1 Intron XP_011543296.1
XM_017017669.1 Intron XP_016873158.1
XM_017017670.1 Intron XP_016873159.1
XM_017017671.1 Intron XP_016873160.1
Gene
MRPL21
Gene Name
mitochondrial ribosomal protein L21
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_181514.1 Intron NP_852615.1
NM_181515.1 Intron NP_852616.1
XM_005273823.4 Intron XP_005273880.1

View Full Product Details