Product Details

SNP ID
rs2069703
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:76617115 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
AGAGTTGTGATATCTTTTCTCCTGC[G/A]TTAACATTTTTAAAATACTGATAAA
Phenotype
MIM: 601919 MIM: 605401
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
F2RL2 PubMed Links

Gene Details

Gene
F2RL2
Gene Name
coagulation factor II thrombin receptor like 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001256566.1 1645 UTR 3 NP_001243495.1
NM_004101.3 1645 UTR 3 NP_004092.1
Gene
IQGAP2
Gene Name
IQ motif containing GTPase activating protein 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001285460.1 1645 Intron NP_001272389.1
NM_001285461.1 1645 Intron NP_001272390.1
NM_001285462.1 1645 Intron NP_001272391.1
NM_006633.3 1645 Intron NP_006624.2
XM_005248410.2 1645 Intron XP_005248467.1
XM_005248414.1 1645 Intron XP_005248471.1
XM_011543108.1 1645 Intron XP_011541410.1
XM_017008960.1 1645 Intron XP_016864449.1
Gene
LOC101929109
Gene Name
uncharacterized LOC101929109
There are no transcripts associated with this gene.

View Full Product Details