Product Details
- SNP ID
-
rs2071182
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.X:15395161 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- AAGTGAGGATAACACTGGCTTGGAG[C/T]GTGAGTGAGTCAGGGTCTCTTGGAG
- Phenotype
-
MIM: 300931
- Polymorphism
- C/T, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
PIR
PubMed Links
Gene Details
- Gene
- PIR
- Gene Name
- pirin
- Gene
- PIR-FIGF
- Gene Name
- PIR-FIGF readthrough
There are no transcripts associated with this gene.
View Full Product Details