Product Details
- SNP ID
-
rs2429440
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.17:46265197 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GATTAACCCACCTCCTTCAGCTGCA[A/G]TTTGGCAAATTTCTCTTCTTTGATT
- Phenotype
-
MIM: 616555
- Polymorphism
- A/G, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
ARL17B
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs112528980] are located under a primer sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- ARL17B
- Gene Name
- ADP ribosylation factor like GTPase 17B
There are no transcripts associated with this gene.
- Gene
- LRRC37A
- Gene Name
- leucine rich repeat containing 37A
View Full Product Details