Product Details
- SNP ID
-
rs2072323
- Assay Type
- Functionally Tested
- NCBI dbSNP Submissions
-
41
- Location
-
Chr.17:9251010 on Build GRCh38
- Set Membership
-
HapMap
JSNP
- Context Sequence [VIC/FAM]
- AATCAGATTATGCATTGATGACGCA[C/T]TGATAAAGCAAAGGGCTTAGGAGAG
- Phenotype
-
MIM: 601614
MIM: 604203
- Polymorphism
- C/T, Transition Substitution
- Allele Nomenclature
-
- Literature Links
-
NTN1
PubMed Links
Gene Details
- Gene
- NTN1
- Gene Name
- netrin 1
There are no transcripts associated with this gene.
- Gene
- STX8
- Gene Name
- syntaxin 8
View Full Product Details