Product Details

SNP ID
rs2240932
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.19:43598858 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CACCTGGGGGCCCCGAAGTGCACCC[A/G]CTGCCTCATCACCTTCGCAGATTCC
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
IRGQ PubMed Links

Gene Details

Gene
IRGQ
Gene Name
immunity related GTPase Q
There are no transcripts associated with this gene.

Gene
ZNF428
Gene Name
zinc finger protein 428
There are no transcripts associated with this gene.

Gene
ZNF576
Gene Name
zinc finger protein 576
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001145347.1 286 Missense Mutation CAC,CGC H38R NP_001138819.1
NM_024327.2 286 Missense Mutation CAC,CGC H38R NP_077303.1

View Full Product Details