Product Details

SNP ID
rs2958522
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.8:144799567 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
AACTTCTGGTTTTGACAAGGGCCTC[A/G]CACAGAATAAACCAAGGAAATAAAA
Phenotype
MIM: 604177
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
MIR6850 PubMed Links

Gene Details

Gene
MIR6850
Gene Name
microRNA 6850
There are no transcripts associated with this gene.

Gene
RPL8
Gene Name
ribosomal protein L8
There are no transcripts associated with this gene.

Gene
ZNF517
Gene Name
zinc finger protein 517
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001317936.1 Intron NP_001304865.1
NM_213605.2 Intron NP_998770.2
XM_011517014.2 Intron XP_011515316.1
XM_011517015.2 Intron XP_011515317.1
XM_011517017.2 Intron XP_011515319.1
XM_017013384.1 Intron XP_016868873.1

View Full Product Details