Product Details

SNP ID
rs2267923
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:42606179 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
TGTTTAGATTTTATAACGTCCTCTT[A/G]TAGCTAAAATTTATTATTGATACTC
Phenotype
MIM: 161565 MIM: 604028
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
LOC101928323 PubMed Links

Gene Details

Gene
LOC101928323
Gene Name
uncharacterized LOC101928323
There are no transcripts associated with this gene.

Gene
NKTR
Gene Name
natural killer cell triggering receptor
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_005385.3 Intron NP_005376.2
XM_005265173.1 Intron XP_005265230.1
XM_006713171.1 Intron XP_006713234.1
XM_006713173.2 Intron XP_006713236.1
XM_011533747.2 Intron XP_011532049.1
XM_011533750.2 Intron XP_011532052.1
XM_017006474.1 Intron XP_016861963.1
XM_017006475.1 Intron XP_016861964.1
XM_017006476.1 Intron XP_016861965.1
XM_017006477.1 Intron XP_016861966.1
XM_017006478.1 Intron XP_016861967.1
XM_017006479.1 Intron XP_016861968.1
XM_017006480.1 Intron XP_016861969.1
Gene
SEC22C
Gene Name
SEC22 homolog C, vesicle trafficking protein
There are no transcripts associated with this gene.

View Full Product Details