Product Details
- SNP ID
-
rs2282065
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.9:97516475 on Build GRCh38
- Set Membership
-
HapMap
- Context Sequence [VIC/FAM]
- TATCCATCCATGGGAAGCAGTTGGA[A/G]CTTGGGAAGCAGTTGGAGCTTAGGA
- Phenotype
-
MIM: 190930
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
TMOD1
PubMed Links
Gene Details
- Gene
- TMOD1
- Gene Name
- tropomodulin 1
View Full Product Details