Product Details

SNP ID
rs2270204
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.9:128280455 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
AGAGGAACCCCAAGCTCTTGAAGTG[G/T]GCTCTGCCTCTTCTCCTTTCTAATA
Phenotype
MIM: 602580 MIM: 616528
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
GOLGA2 PubMed Links

Gene Details

Gene
GOLGA2
Gene Name
golgin A2
There are no transcripts associated with this gene.

Gene
SWI5
Gene Name
SWI5 homologous recombination repair protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001040011.1 Intron NP_001035100.1
NM_001318089.1 Intron NP_001305018.1
NM_001318092.1 Intron NP_001305021.1
XM_011518652.2 Intron XP_011516954.1

View Full Product Details