Product Details

SNP ID
rs2235091
Assay Type
Validated
NCBI dbSNP Submissions
NA
Location
Chr.19:50907215 on Build GRCh38
Set Membership
HapMap Validated
Context Sequence [VIC/FAM]
ATCATCAATAACAACATAGCCCAGC[G/A]ACATATCTGTTGGAAGTCCCATTCC
Phenotype
MIM: 603767
Polymorphism
G/A, Transition substitution
Allele Nomenclature
Literature Links
KLK4 PubMed Links

Gene Details

Gene
KLK4
Gene Name
kallikrein related peptidase 4
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001302961.1 Intron NP_001289890.1
NM_004917.4 Intron NP_004908.4
XM_005259441.4 Intron XP_005259498.2
XM_011527545.2 Intron XP_011525847.1
XM_011527546.2 Intron XP_011525848.1

View Full Product Details