Product Details

SNP ID
rs2289047
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.13:109755468 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GAAGTACCTACATAAACATCTACAT[A/C]GAGAAGATTAAACAAGTCTGTTAAA
Phenotype
MIM: 600797
Polymorphism
A/C, Transversion substitution
Allele Nomenclature
Literature Links
IRS2 PubMed Links

Gene Details

Gene
IRS2
Gene Name
insulin receptor substrate 2
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_003749.2 5367 UTR 3 NP_003740.2

View Full Product Details