Product Details

SNP ID
rs2289289
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.5:10683303 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
TCTGCGCTTAAGTCACAATGTTGGG[A/T]CTCACCAGACGGCGGCTCTGGAAAG
Phenotype
MIM: 600954
Polymorphism
A/T, Transversion substitution
Allele Nomenclature
Literature Links
DAP PubMed Links

Gene Details

Gene
DAP
Gene Name
death associated protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291963.1 Intron NP_001278892.1
NM_004394.2 Intron NP_004385.1

View Full Product Details