Product Details

SNP ID
rs2276318
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.11:65890442 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
GGAGGTTCGAAGCTCCTAGTTTCGG[A/G]TTGAGACCGATGTGGAGGGCGATGA
Phenotype
MIM: 605360 MIM: 602364 MIM: 608296 MIM: 136515
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
CCDC85B PubMed Links

Gene Details

Gene
CCDC85B
Gene Name
coiled-coil domain containing 85B
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_006848.2 39 UTR 5 NP_006839.2
Gene
CTSW
Gene Name
cathepsin W
There are no transcripts associated with this gene.

Gene
FIBP
Gene Name
FGF1 intracellular binding protein
There are no transcripts associated with this gene.

Gene
FOSL1
Gene Name
FOS like 1, AP-1 transcription factor subunit
There are no transcripts associated with this gene.

View Full Product Details