Product Details

SNP ID
rs2969071
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.7:2473807 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
GATTGATGTTGATGACAATGATGAC[A/G]GATATAGATGGTATATACATGATAG
Phenotype
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
GRIFIN PubMed Links

Gene Details

Gene
GRIFIN
Gene Name
galectin-related inter-fiber protein
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_001291784.1 1957 Intron NP_001278713.1
XM_011515405.2 1957 UTR 3 XP_011513707.1

View Full Product Details