Transcript Accession | SNP Location | SNP Type | Codon Change | Amino Acid Change | Protein ID |
---|---|---|---|---|---|
NM_001001895.2 | 124 | Missense Mutation | CTC,TTC | L28F | NP_001001895.1 |
NM_001243467.1 | 124 | Missense Mutation | CTC,TTC | L28F | NP_001230396.1 |
NM_018961.3 | 124 | Missense Mutation | CTC,TTC | L28F | NP_061834.1 |
XM_006724013.3 | 124 | UTR 5 | XP_006724076.1 | ||
XM_011529605.2 | 124 | Missense Mutation | CTC,TTC | L28F | XP_011527907.1 |
XM_011529606.2 | 124 | UTR 5 | XP_011527908.1 | ||
XM_011529607.2 | 124 | UTR 5 | XP_011527909.1 | ||
XM_011529609.1 | 124 | Missense Mutation | CTC,TTC | L28F | XP_011527911.1 |
XM_011529610.2 | 124 | Intron | XP_011527912.1 |