Product Details
- SNP ID
-
rs2175133
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.2:207714623 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GAATGCACTGGACAGAGGAATATTA[A/G]AAGCTGGCAAGAGGGAAGGTAAGGT
- Phenotype
-
- Polymorphism
- A/G, Transition substitution
- Allele Nomenclature
-
- Literature Links
-
CCNYL1
PubMed Links
Gene Details
- Gene
- CCNYL1
- Gene Name
- cyclin Y like 1
View Full Product Details