Product Details

SNP ID
rs2597771
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.4:17489747 on Build GRCh38
Set Membership
HapMap
Context Sequence [VIC/FAM]
CATAAAAAGTGTTCTGTAAGTACAC[A/G]TTTCAAACATAAAACGCAAGTCCCA
Phenotype
MIM: 612676
Polymorphism
A/G, Transition substitution
Allele Nomenclature
Literature Links
QDPR PubMed Links

Gene Details

Gene
QDPR
Gene Name
quinoid dihydropteridine reductase
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_000320.2 Intron NP_000311.2
NM_001306140.1 Intron NP_001293069.1

View Full Product Details