Product Details
- SNP ID
-
rs2683596
- Assay Type
- Functionally tested
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.10:124763495 on Build GRCh38
- Set Membership
-
- Context Sequence [VIC/FAM]
- GCAAGGCAGTGTATTCTGTTACTGT[C/G]CCATATCTAACTCCAAATACCATTT
- Phenotype
-
- Polymorphism
- C/G, Transversion substitution
- Allele Nomenclature
-
- Literature Links
-
METTL10
PubMed Links
Gene Details
- Gene
- METTL10
- Gene Name
- methyltransferase like 10
View Full Product Details