Product Details
- SNP ID
-
rs2657880
- Assay Type
- Validated
- NCBI dbSNP Submissions
-
NA
- Location
-
Chr.12:56469986 on Build GRCh38
- Set Membership
-
HapMap
Validated
- Context Sequence [VIC/FAM]
- ACTTGTTCTTGGGGCTCTACCAGAT[C/G]GCTGAAGAGTAAATCCTTTCTACCT
- Phenotype
-
MIM: 606365
- Polymorphism
- C/G, Transversion Substitution
- Allele Nomenclature
-
- Literature Links
-
GLS2
PubMed Links
- Additional Information
-
For this assay, SNP(s) [rs74811816] are located under a probe sequence. To help evaluate the possible impact of a given SNP on your experiment, please refer to the 1000 Genomes and NCBI dbSNP databases. A higher minor allele frequency in your study population represents a higher risk to assay performance.
Gene Details
- Gene
- GLS2
- Gene Name
- glutaminase 2
- Gene
- SPRYD4
- Gene Name
- SPRY domain containing 4
Transcript Accession |
SNP Location |
SNP Type |
Codon Change |
Amino Acid Change |
Protein ID |
NM_207344.3 |
1108 |
UTR 3 |
|
|
NP_997227.1 |
View Full Product Details