Product Details

SNP ID
hCV15930015
Assay Type
Functionally tested
NCBI dbSNP Submissions
NA
Location
Chr.3:49688282 on Build GRCh38
Set Membership
Context Sequence [VIC/FAM]
CCTGGCTAGGCATTCAGACTCCAAA[G/T]CTGGGCTCTCACCTGCACAAAGGCA
Phenotype
MIM: 102645 MIM: 142408 MIM: 614472
Polymorphism
G/T, Transversion substitution
Allele Nomenclature
Literature Links
APEH PubMed Links

Gene Details

Gene
APEH
Gene Name
acylaminoacyl-peptide hydrolase
There are no transcripts associated with this gene.

Gene
MST1
Gene Name
macrophage stimulating 1
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_020998.3 Intron NP_066278.3
Gene
RNF123
Gene Name
ring finger protein 123
Transcript Accession SNP Location SNP Type Codon Change Amino Acid Change Protein ID
NM_022064.4 Intron NP_071347.2

View Full Product Details